Order Entry
Belgium
ContactUsLinkComponent
Anti-Transglutaminase 2 Mouse Monoclonal Antibody [clone: CUB 7402]
Anti-Transglutaminase 2 Mouse Monoclonal Antibody [clone: CUB 7402]
Catalog # ABCAAB2386-500
Supplier:  Abcam
undefined
Anti-Transglutaminase 2 Mouse Monoclonal Antibody [clone: CUB 7402]
Catalog # ABCAAB2386-500
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    transglutaminase 2
  • Antigen symbol:
    TGM2
  • Clonality:
    Monoclonal
  • Clone:
    CUB 7402
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    1418
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pack type:
    Vial
  • Pk:
    500 µl

Specifications

About this item

Mouse monoclonal [CUB 7402] to Transglutaminase 2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Transglutaminase 2
Clonality: Monoclonal
Clone: CUB 7402
Conjugation:
Epitope: Epitope mapping for this antibody is amino acid 447-478
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Guinea pig;Dog;Cat;Rabbit