Order Entry
Belgium
ContactUsLinkComponent
Anti-SOD1 Rabbit Polyclonal Antibody
Anti-SOD1 Rabbit Polyclonal Antibody
Catalog # BIRBORB95892-50
Supplier:  Biorbyt
undefined
Anti-SOD1 Rabbit Polyclonal Antibody
Catalog # BIRBORB95892-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Superoxide Dismutase 1
  • Antigen symbol:
    SOD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage buffer:
    PBS, pH 7.0, 0,1% sodium azide, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Immunochemistry: 10 ?g/ml: 0.2 ?g/ml

Type: Primary
Antigen: SOD1 (superoxide dismutase 1, soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: