Anti-E7 protein of human Papillomavirus type 18 Mouse Monoclonal Antibody
Catalogus # BIRBORB79454-1
Leverancier: Biorbyt
Specificaties
- Antibody type:Primary
- Antigen name:E7 Protein of human Papillomavirus type 18
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 mg
- Pk:1 mg
Specificaties
Over dit artikel
Type: Primary
Antigen: E7 protein of human Papillomavirus type 18
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human