Anti-SH3 domain binding protein 1 Rabbit Polyclonal Antibody
N° de catalogue BIRBORB73594-100
Fournisseur: Biorbyt
Spécifications
- Antibody type:Primary
- Antigen name:SH3 domain binding Protein 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Spécifications
À propos de cet article
Type: Primary
Antigen: SH3 domain binding protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human