Anti-OSBPL1A Rabbit Polyclonal Antibody
BIRBORB73593-100
: Biorbyt
- Antibody type:Primary
- Antigen name:oxysterol binding Protein-like 1A
- Antigen symbol:OSBPL1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: OSBPL1A (oxysterol binding protein-like 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human