Anti-SLC18A3 Mouse Monoclonal Antibody
Catalog # BIRBORB67481-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 18 (vesicular acetylcholine transporter), member 3
- Antigen symbol:SLC18A3
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide amino acids 521-532 of human VACht
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunochemistry: 1:50-1:200
Type: Primary
Antigen: SLC18A3 (solute carrier family 18 (vesicular acetylcholine transporter), member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat