Anti-NLRP6 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BIRBORB123790-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:NLR family, pyrin domain containing 6
- Antigen symbol:NLRP6
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:99 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NLRP6 (NLR family, pyrin domain containing 6)
Clonality: polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: