Order Entry
Belgium
ContactUsLinkComponent
Anti-Proximal Nephrogenic Renal Antigen, cln PN-15 Mouse Monoclonal Antibody
Anti-Proximal Nephrogenic Renal Antigen, cln PN-15 Mouse Monoclonal Antibody
Catalog # BIRBORB108788-100
Supplier:  Biorbyt
Anti-Proximal Nephrogenic Renal Antigen, cln PN-15 Mouse Monoclonal Antibody
Catalog # BIRBORB108788-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proximal Nephrogenic Renal Antigen, cln PN-15
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with renal cortical tissue extract and mouse myeloma cells
  • Purification:
    purified by Protein A/G Chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Proximal Nephrogenic Renal Antigen, cln PN-15
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human