Anti-Aflatoxin B1/M1 Rat Monoclonal Antibody
Catalog # BIRBORB107993-0.5
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Aflatoxin B1/M1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG
- Form:lyophilized
- Storage buffer:0,01 M PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:0,5 mg/vial
- Shipping temperature:Ice
- Immunogen:Aflatoxin M1-BSA Conjugate
- Purification:purified by affinity chromatography
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Primary
Antigen: Aflatoxin B1/M1 (aflatoxin B1/M1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: