Anti-P2RY12 Rabbit Polyclonal Antibody
Catalog # BIRBORB107569-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:purinergic receptor P2Y, G-protein coupled, 12
- Antigen symbol:P2RY12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: P2RY12 (purinergic receptor P2Y, G-protein coupled, 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat