Anti-EC SOD Rabbit Polyclonal Antibody Atto Dyes and Enzyme-Protein Labels
Catalog # BIRBORB107266-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:EC SOD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:Phosphate-buffered solution 0,09% sodium azide and 0,2% (w/v) BSA
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Peptide corresponding to AA 227-236 of human EC SOD
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EC SOD
Clonality: polyclonal
Clone:
Conjugation: Atto Dyes and Enzyme-Protein Labels
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat