Order Entry
Belgium
ContactUsLinkComponent
Mouse IgG2a Isotype Control (Unconjugated)
Mouse IgG2a Isotype Control (Unconjugated)
Catalog # BIRBORB43542-0.1
Supplier:  Biorbyt
Mouse IgG2a Isotype Control (Unconjugated)
Catalog # BIRBORB43542-0.1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Control
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Isotype:
    IgG2a
  • Storage buffer:
    Azide Free.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Control
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG2a
Reactivity: