Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mouse IgG2a Isotype Control (Unconjugated)

Mouse IgG2a Isotype Control (Unconjugated)

100 μg
Catalog # BIRBORB43541-0.1
Marketsource
Supplier:   Biorbyt
Mouse IgG2a Isotype Control (Unconjugated)
Catalog # BIRBORB43541-0.1
Supplier:  Biorbyt
Marketsource
Mouse IgG2a Isotype Control (Unconjugated)

100 μg

Price
Inquire For Price
Log in for availability
1 * 0,1 mg
Product Details & Documents

Type: Control
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG2a
Reactivity:

Documents

Specifications
  • Size:
    100 μg
  • Cat. No.:
    BIRBORB43541-0.1
  • Flow cytometry:
    Yes
  • Purification:
    purified
  • Antibody type:
    Control
  • Isotype:
    IgG2a
  • Storage temperature:
    Cool
  • Conjugation:
    Unconjugated
  • Shipping temperature:
    Ice
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.