Anti-ASB4 Rabbit Polyclonal Antibody
100 µl
Catalog # BIRBORB41338-100
Marketsource
Supplier: Biorbyt
Jump to:
Product Details & Documents
Western Blot: 1:500-1:3000
Type: Primary
Antigen: ASB4 (ankyrin repeat and SOCS box containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Size:100 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by antigen affinity chromatography
- Antibody type:Primary
- Amino acid number:29 to 327
- Storage temperature:Cool
- Host:Rabbit
- Concentration:0,74 mg/ml
- Shipping temperature:Ice
- Antigen name:ankyrin repeat and SOCS box containing 4
- Cat. No.:BIRBORB41338-100
- Reactivity:Human
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 29 and 327 of Human ASB4.
- Conjugation:Unconjugated
- Antigen symbol:ASB4
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



