Anti-CDK1 Rabbit Polyclonal Antibody
Catalogus # BIRBORB41337-100
Leverancier: Biorbyt
Specificaties
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1 to 280
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 280 of Human TBRG4.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specificaties
Over dit artikel
Western Blot: 1:500-1:3000
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human