Anti-Basal Cellmarker Cocktail p63 Protein + HMW Cytokeratin Mouse monoclonal antibody
Catalog # BIRBORB27390-5
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Basal Cellmarker Cocktail p63 Protein + HMW Cytokeratin
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a,IgG1
- Reactivity:Human
- Amino acid number:1 to 205
- Storage temperature:Cool
- Concentration:neat
- Shipping temperature:Ice
- Immunogen:Recombinant human p63 protein (aa 1-205) and cytokeratin extract of human stratum corneum
- Size:5 ml
- Pk:5 mL
Specifications
About this item
Type: Primary
Antigen: Basal Cellmarker Cocktail p63 Protein + HMW Cytokeratin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1+IgG2a
Reactivity: Human