Order Entry
Belgium
ContactUsLinkComponent
Anti-APOB Rabbit Polyclonal Antibody
Anti-APOB Rabbit Polyclonal Antibody
Catalog # BIRBORB25178-1
Supplier:  Biorbyt
Anti-APOB Rabbit Polyclonal Antibody
Catalog # BIRBORB25178-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein B
  • Antigen symbol:
    APOB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Amino acid number:
    48 to 100
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse low density lipoprotein (LDL).
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse