Order Entry
Belgium
ContactUsLinkComponent
Anti-Rat Guanylin Rabbit Polyclonal Antibody
Anti-Rat Guanylin Rabbit Polyclonal Antibody
Catalog # BIRBORB24208-20
Supplier:  Biorbyt
Anti-Rat Guanylin Rabbit Polyclonal Antibody
Catalog # BIRBORB24208-20
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Rat Guanylin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Form:
    lyophilized
  • Amino acid number:
    78 to 92
  • Sequence:
    PNTCEICAYAACTGC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic rat Guanylin (a.a. 78-92) (PNTCEICAYAACTGC).
  • Size:
    20 µl
  • Pk:
    20 µl

Specifications

About this item

ELISA: 1:3000

Type: Primary
Antigen: Rat Guanylin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat