Anti-Sialoprotein Rabbit Polyclonal Antibody
BIRBORB24185-20
: Biorbyt
- Antibody type:Primary
- Antigen name:Sialoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Form:lyophilized
- Amino acid number:262 to 279
- Sequence:YESENGEPRGDNYRAYED
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic human Bone Sialoprotein peptide (YESENGEPRGDNYRAYED).
- Size:20 µl
- Pk:20 µl
ELISA: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: Sialoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human