Anti-E. coli Enterotoxin B Mouse Monoclonal Antibody
BIRBORB23992-1
: Biorbyt
- Antibody type:Primary
- Antigen name:E. coli Enterotoxin B
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:5,5 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Recombinant E. coli heat-labile Enterotoxin B-chain (LTB), full-length.
- Purification:purified by salt precipitation
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: E. coli Enterotoxin B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: