Anti-Herpes simplex Type-2 Sheep Polyclonal Antibody (FITC (Fluorescein))
BIRBORB23095-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Herpes simplex Type-2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Sheep
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage buffer:0,01 M PBS, pH 7,2 containing 10 mg/mL BSA.
- Storage temperature:Cool
- Concentration:4-5 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:HSV Type 2, strain G (Human).
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Herpes simplex Type-2
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: none
Reactivity: