Order Entry
Belgium
ContactUsLinkComponent
Anti-E. coli (all antigens) Goat Polyclonal Antibody (FITC (Fluorescein))
Anti-E. coli (all antigens) Goat Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB23085-1
Supplier:  Biorbyt
Anti-E. coli (all antigens) Goat Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB23085-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    E. coli (all antigens)
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Storage buffer:
    0,01M PBS, pH 7,2 containing 10mg/ml BSA
  • Storage temperature:
    Cool
  • Concentration:
    4-5 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Mixture of E. coli serotypes
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: E. coli (all antigens)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: