Anti-IgG1 Rabbit Polyclonal Antibody
Catalog # BIRBORB22400-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Pools of purified homogenous IgG1 isolated from rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: Rat IgG1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: