Order Entry
Belgium
ContactUsLinkComponent
 
Anti-IgA/IgG1/IgG2a/IgG2b/IgG3/IgM Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BIRBORB22099-1
 :  Biorbyt
Anti-IgA/IgG1/IgG2a/IgG2b/IgG3/IgM Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BIRBORB22099-1
 :  Biorbyt

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgA/IgG1/IgG2a/IgG2b/IgG3/IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • Format:
    immunoconjugate
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Concentration:
    7,2 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,6, No foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified polyclonal Mouse IgG, and homogenous IgA and IgM isolated from mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3 IgA IgM
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: