Order Entry
Belgium
ContactUsLinkComponent
Anti-IgA/IgG1/IgG2a/IgG2b/IgG2c/IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgA/IgG1/IgG2a/IgG2b/IgG2c/IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB22089-1
Supplier:  Biorbyt
Anti-IgA/IgG1/IgG2a/IgG2b/IgG2c/IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB22089-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgA/IgG1/IgG2a/IgG2b/IgG2c/IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Goat
  • Format:
    immunoconjugate
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,7, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified polyclonal rat IgG, and homogenous IgA and IgM isolated from rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: Rat IgG1 IgG2A IgG2B IgG2C IgA IgM
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: