Anti-lambda light chain Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21910-1
Supplier: Biorbyt
Specifications
- Type d'anticorps:Secondaire
- Nom de l'antigène:lambda light chain
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Goat
- Format:immunoconjugate
- Température de stockage:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein ratio (F/P) approximately 1,6, no foreign proteins added,
- Température de transport:Ice
- Immunogène:A pool of purified Bence Jones lambda proteins isolated from human urine. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Taille:1 ml
- Cdt:1 mL
Specifications
About this item
Type: Secondary
Antigen: lambda light chain
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: none
Reactivity: