Anti-IgG1/IgG2a/IgG2b/IgG2c Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB21695-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG2c
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Peroxidase/IgG protein molar ratio (E/P) is approximately 1,6, no foreign proteins added
- Shipping temperature:Ice
- Immunogen:Purified normal IgG, including all known subclasses, isolated from pooled rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: Rat IgG1 IgG2A IgG2B IgG2C
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: