Order Entry
Belgium
ContactUsLinkComponent
Anti-IgG1/IgG2a/IgG2b/IgG2c Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG1/IgG2a/IgG2b/IgG2c Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB21545-1
Supplier:  Biorbyt
Anti-IgG1/IgG2a/IgG2b/IgG2c Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB21545-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG1/IgG2a/IgG2b/IgG2c
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • Format:
    immunoconjugate
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, Enzyme/IgG protein molar ratio (E/P) is approximately 1,5, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified normal IgG isolated from pooled rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: Rat IgG1 IgG2A IgG2B IgG2C
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: