Anti-Kappa Light Chainkappa light chain Rabbit Polyclonal Antibody (Biotin)
Catalog # BIRBORB21519-10
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Biotin/IgG protein molar ratio (B/P) is approximately 4,4, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:A pool of purified Bence Jones kappa proteins isolated from human urine. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: