Anti-IgM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB21362-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Peroxidase/IgG protein molar ratio (E/P) is approximately 1,5, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified homogenous IgM isolated from pooled mouse serum. Immunization with intact (19S) and split IgM (7S). Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: