Anti-L-Lactic Dehydrogenase Rabbit Polyclonal Antibody
BIRBORB20786-1
: Biorbyt
- Antibody type:Primary
- Antigen name:L-Lactic Dehydrogenase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig
- Form:lyophilized
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:1 mg/0,5ml, no foreign proteins added
- Shipping temperature:Ice
- Immunogen:L-Lactic dehydrogenase isolated and purified from porcine heart. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: L-Lactic Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pig