Anti-E2F7 Goat Polyclonal Antibody
Catalog # BIRBORB20204-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:E2F transcription factor 7
- Antigen symbol:E2F7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:90 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: E2F7 (E2F transcription factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human