Anti-Neurofibromin 2 Goat Polyclonal Antibody
BIRBORB19615-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Neurofibromin 2
- Antigen symbol:NF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Zebrafish
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA._x000D_
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide (Internal Region)
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Neurofibromin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Zebrafish