Order Entry
Belgium
ContactUsLinkComponent
Anti-FOXD1 Goat Polyclonal Antibody
Anti-FOXD1 Goat Polyclonal Antibody
  BIRBORB18553-100
 :  Biorbyt
Anti-FOXD1 Goat Polyclonal Antibody
  BIRBORB18553-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Forkhead Box D1
  • Antigen symbol:
    FOXD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    C Terminus
  • Purification:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

ELISA: 1:16000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: FOXD1 (forkhead box D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat