Order Entry
Belgium
ContactUsLinkComponent
 
Anti-GRGDS (Gly-Arg-Gly-Asp-Ser) Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB15695-100
Supplier:  Biorbyt
Anti-GRGDS (Gly-Arg-Gly-Asp-Ser) Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB15695-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Gly-Arg-Gly-Asp-Ser
  • Antigen symbol:
    GRGDS
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: GRGDS (Gly-Arg-Gly-Asp-Ser)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: