Order Entry
Belgium
ContactUsLinkComponent
Anti-PI3K P85 alpha/ gamma / beta Rabbit Polyclonal Antibody
Anti-PI3K P85 alpha/ gamma / beta Rabbit Polyclonal Antibody
Catalog # BIRBORB14711-100
Supplier:  Biorbyt
undefined
Anti-PI3K P85 alpha/ gamma / beta Rabbit Polyclonal Antibody
Catalog # BIRBORB14711-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PI3K P85 alpha/gamma/beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Storage buffer:
    PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    The antiserum was produced against synthesized non-phosphopeptide derived from human PI3K P85 alpha/gamma/beta around the phosphorylation site of tyrosine 467/199/464 (L -YP-E-E-Y).
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PI3K P85 alpha/ gamma / beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat