Order Entry
Belgium
ContactUsLinkComponent
Anti-c-Myc tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-c-Myc tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB13897-100
Supplier:  Biorbyt
Anti-c-Myc tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB13897-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EQKLISEEDL
  • Antigen symbol:
    c-Myc tag
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    49 kDa
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: c-Myc tag (c-Myc tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human