Anti-Hemagglutinin/Influenza A virus (H5N1) Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB9032-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Hemagglutinin/Influenza A virus
- Antigen symbol:H5N1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human Anhti-Hemagglutinin/Influenza A virus (H5N1).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: Hemagglutinin/Influenza A virus (H5N1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus