Anti-MAGEB1 Mouse Monoclonal Antibody [clone: OTI5C12]
Catalog # ORIGTA700359
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Melanoma Antigen Family B1
- Antigen symbol:MAGEB1
- Clonality:Monoclonal
- Clone:OTI5C12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) with 0,05% sodium azide, 10mg/ml BSA, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: MAGEB1 (melanoma antigen family B, 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human