Anti-CDC25A Rabbit Polyclonal Antibody
Catalog # ORIGTA313589
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Cell division cycle 25A
- Antigen symbol:CDC25A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Modification:phospho-Thr507
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: