Order Entry
Belgium
ContactUsLinkComponent
Anti-MAP2K6 Rabbit Polyclonal Antibody
Anti-MAP2K6 Rabbit Polyclonal Antibody
  ORIGTA309987
 :  OriGene
Anti-MAP2K6 Rabbit Polyclonal Antibody
  ORIGTA309987
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase kinase 6
  • Antigen symbol:
    MAP2K6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Modification:
    phospho-Ser207
  • Storage buffer:
    phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    The antibody was purified by affinity chromatography from rabbit antiserum by affinity-chromatography usingepitope-specific phosphopeptide. The antibody against non-phosphopeptide was removedby chromatogramphy using non-phosphopeptide corresponding to the phosphorylation site.
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat