Order Entry
Belgium
ContactUsLinkComponent
Rat IgG2b Isotype Control [clone: LTF-2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Rat IgG2b Isotype Control [clone: LTF-2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # 65-4031-U100
Supplier:  Tonbo Biosciences
Rat IgG2b Isotype Control [clone: LTF-2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # 65-4031-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    LTF-2
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: IgG2b (Rat) (rat IgG2b)
Clonality: monoclonal
Clone: LTF-2
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: