Anti-RPS6KB2 Rabbit Polyclonal Antibody
ABGEAP8009C
: Abgent
- Antibody type:Primary
- Antigen name:Ribosomal Protein S6 Kinase, 70kDa, Polypeptide 2
- Antigen symbol:RPS6KB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Gene ID:3061
- Antigen synonyms:p70 ribosomal S6 kinase beta|70 kDa ribosomal protein S6 kinase 2|p70(S6K)-beta|P70-beta|S6K2|SRK|Ribosomal protein S6 kinase beta 2|KLS|p70S6Kb|S6K-beta2|P70-beta-1|P70-beta-2|STK14B|Serine/threonine-protein kinase 14 beta|S6 kinase-related kinase
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:AEVKQMRARRKTAKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This HCRTR1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 270-298 amino acids from the Central region of human HCRTR1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat