Anti-KDM3B Rabbit Polyclonal Antibody
ABGEAP2312B
: Abgent
- Antibody type:Primary
- Antigen name:ADP-Ribosyltransferase 3
- Antigen symbol:KDM3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:ART3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ART3 (ADP-ribosyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human