Anti-POMC Rabbit Polyclonal Antibody
Catalog # ABGEAP2311A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:ADP-Ribosyltransferase 1
- Antigen symbol:POMC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal ( 1–50 aa)
- Cross adsorption:No
- Antigen synonyms:ART1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ART1 (ADP-ribosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse