Anti-ORML2 Rabbit Polyclonal Antibody
ABGEAP10815C
: Abgent
- Antibody type:Primary
- Antigen name:ORMDL Sphingolipid Biosynthesis Regulator 2
- Antigen symbol:ORMDL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (1021–1035 aa)
- Cross adsorption:No
- Antigen synonyms:Tyrosine-protein kinase receptor EHK-2|Ephrin type-A receptor 6|EPH homology kinase-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ORML2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse