Order Entry
Belgium
ContactUsLinkComponent
Cleanroom wipes, Kimtech™ Pure W4
Cleanroom wipes, Kimtech™ Pure W4
Catalog # KIMB33330
Supplier:  KIMBERLY CLARK
undefined
Cleanroom wipes, Kimtech™ Pure W4
Catalog # KIMB33330
Supplier:  KIMBERLY CLARK

Specifications

  • Packed:
    100 wipes/pack, 5 packs/case
  • Description:
    W4
  • L×W:
    305×305 mm
  • Seq:
    02
  • Model:
    W4
  • Pk:
    500

Specifications

About this item

These critical task wipes are made of 100% PP and acid, base and solvent-resistant.

  • Thermally bonded with no glues or binders
  • Straight, clean, knife-cut edges minimise lint
  • Meet the U.S. FDA requirements for food packaging materials

Ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries.

Suitable for use in Class 10, ISO 4, M2.5 cleanrooms.

Colour: White

Packaging: Double-bagged in anti-stat packaging.