ContactUsLinkComponent
Adapters with screw thread and standard gound joint cone
Adapters with screw thread and standard gound joint cone
Catalog # QVFLVBS0101404
Supplier:  QVF LABORTECHNIK
Adapters with screw thread and standard gound joint cone
Catalog # QVFLVBS0101404
Supplier:  QVF LABORTECHNIK
Specifications
  • Cone size:
    29/32 NS
  • Hole Ø:
    8,8 - 9,3 mm
  • Thread:
    GL 18
  • Pk:
    1
Specifications
About this item
Connecters, ground cone with thread and screw cap, can be used to assemble the thermometers without ground joint. The seal is part of delivery.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes