ContactUsLinkComponent
Scraper, SP Bel-Art
Scraper, SP Bel-Art
Catalog # BELAH368200000
undefined
Scraper, SP Bel-Art
Catalog # BELAH368200000
Specifications
  • Wiping area app.:
    140×63 mm
  • Length:
    286 mm
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Polyamide, glass-reinforced. Rigid and strong one piece scraper for heavy use.
  • Flat blade with chisel edge
  • Blade may be resharpened to restore chisel edge