ContactUsLinkComponent
Sodium hydroxide 8 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide 8 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
  4.80763.9200
 :  
Sodium hydroxide 8 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
  4.80763.9200
 :  
  • Pk:
    200 L
  • Seq:
    5549
 :
 :40 g/mol
 :109 °C (1013 hPa)
 :≤1.11 g/cm³ (20 °C)
 :Ambient
 :215-185-5
 :1824
 :8,II
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548